Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury Metal, Instrument Grade, Reagents

Catalog No. 01337179
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-179 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-179 Supplier Reagents Holdings Llc Supplier No. C2244000100A6
Only null left
Add to Cart
Add to Cart

Mercury Metal, Instrument Grade, Instrument, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7439-97-6
Molecular Formula Hg
Molecular Weight (g/mol) 200.59
MDL Number MFCD00011035
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
Synonym Hg
IUPAC Name mercury
SMILES [Hg]

Specifications

Color Silver-White
Melting Point -38.87°C
Boiling Point 356.6°C
Avg. Mol. Wt. or Mol. Wt. Range 200.59
Quantity 100 mL
Packaging Poly Bottle
Form Liquid
Chemical Name or Material Mercury Metal, Instrument Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.