Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury, plasma standard solution, Specpure™ Hg 1000μg/mL

Catalog No. AA13865AE
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA13865AE 100 mL
AA13865AD 50 mL
AA13865AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA13865AE Supplier Thermo Scientific Chemicals Supplier No. 013865AE
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7439-97-6 | Hg | 200.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-97-6 , 7697-37-2 , 7732-18-5
Molecular Formula Hg
Molecular Weight (g/mol) 200.59
MDL Number MFCD00011035
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
PubChem CID 23931
ChEBI CHEBI:16170
IUPAC Name mercury
SMILES [Hg]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
Type Analytical Standard
Quantity 100 mL
Grade Specpure
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Mercury: 0.1%; Water: 94.9%
Chemical Name or Material Mercury
Hazard Category H290-H314-H335-H373
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.