Learn More
New Pig Mercury Spill Kit in Bucket
SDP
Catalog No. 18389128 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Mercury Spill Kit in Bucket
Catalog No.
18-389-128 Type: Mercury Spill Kit in Bucket
1 options
This item is not returnable. View return policy
Catalog No. 18-389-128 Supplier New Pig Supplier No. KIT600
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safely clean up mercury spills and suppress harmful vapors with this specially equipped kit containing absorbents, wipers and PPE.
  • Includes PIG Mercury Absorbent Powder to transform up to 1000 grams of visible mercury into a zinc amalgam
  • Suppress hazardous mercury vapors to limit worker exposure
  • Sweep up stabilized mercury with hand broom and dust pan for easy transfer into disposal bags (all components included)
  • Lightweight, 100% polyethylene container resists chemicals Screw-on lid opens easily for access to supplies
  • Includes PPE to guard against hazardous materials
  • Contents are organized for quick, efficient response
  • Use PIG Mercury Cleanup Wipers to decontaminate hands, tools and surfaces
Specifications
Material Polypropylene