Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ MERIFLUOR™ VZV Direct Fluorescent Test Kit
GSA_VA

Catalog No. 14910114 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
40 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-114 40 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-114 Supplier Meridian Bioscience™ Supplier No. 500113
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Direct fluorescent antibody detection of Varicella-Zoster virus in vesicle smears, biopsies, VZV cell culture

  • Provides specific differentiation of VZV from Herpes Simplex Virus or Cytomegalovirus
  • Swab specimen
  • Assay format: IFA
  • Shelf life: 15 months

Certifications

CE marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Detectable Analytes Varicella Zoster Virus
Format IFA
Type MeriFluor™ VZV
For Use With (Application) Direct fluorescent Antibody detection of Varicella-Zoster virus in vesicle smears, biopsies, VZV cell culture
Quantity 40 Tests
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.