Learn More
Meridian Bioscience™ Para-Pak™ EcoFix™ Stool Transportation and Fixative Systems
GSA_VA
Catalog No. 14910170 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
120/Cs.
200/Cs.
84/Cs.
Media Type:
EcoFix Fixative
EcoFix Fixative, C&S, Clean
LV-PVA, 10% Formalin, Clean
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
14-910-170 84/Cs. EcoFix Fixative, C&S, Clean
22-045274 120/Cs. LV-PVA, 10% Formalin, Clean
23-043095 120/Cs. EcoFix Fixative
14-910-167 200/Cs. EcoFix Fixative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-910-170 Supplier Meridian Bioscience™ Supplier No. 331312
Add to Cart
Add to Cart
Includes
A spork for specimen manipulation, a squeezable vial for rapid specimen filtration and a modified cap that adapts to 15- or 50mL conical processing tubes
Formalin-free fixative
  • Designed for collecting, transporting, filtering, preserving and examining stool specimens for intestinal parasites
  • Works with trichrome and iron hematoxylin stain procedures
  • Preservative eliminates the need for processing in a hood, reduces disposal cost and minimizes the adverse effects from delay in specimen transport
  • Ideal for the recovery of eggs and cysts in concentration procedures
  • Both concentration and permanent stain may be performed on the EcoFix-preserved specimen
  • For each of the systems, there is also a corresponding clean system kit
EcoFix Fixative System A single vial containing EcoFix zinc-based preservative designed for collecting, transporting, filtering and preserving fecal parasites EcoFix Fixative/Clean Vial System A two-vial system including an EcoFix vial for processing preserved fecal specimens and a leakproof, 30mL clean vial for transporting specimens that do not need a preservative transport media
EcoFix ULTRA System Combines the concentration benefits of the Para-Pak ULTRA with the formalin-free EcoFix fixative. In the ULTRA format, a filter is incorporated into the vial cap.
EcoFix ULTRA/Clean Vial System Two-vial system includes both an EcoFix ULTRA vial (described above) for processing preserved fecal specimens and a clean vial for processing fecal specimens that have not been preserved.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Stool Transport System
Certifications/Compliance CE
Includes Built-in concentrator, spork for specimen manipulation, a squeezable vial for rapid specimen filtration and a modified cap that adapts to 15 or 50mL conical processing tubes
Media Type EcoFix Fixative, C&S, Clean
No. of Vials 3
Quantity 84/Cs.
Storage Requirements 20°C to 30°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov