Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Premier™ Platinum HpSA™ Plus
GSA_VA

Catalog No. 23315085 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-085 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-315-085 Supplier Meridian Bioscience™ Supplier No. 601396
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Antibody-coated microwells; positive control; negative control; sample diluent; 20X wash buffer; enzyme conjugate; substrate; stop solution; transfer pipets; microwell strip holder; microwell strip sealer; and wooden stick applicators

Requires

Test tubes (for dilution of sample), distilled water, squirt bottle, graduated cylinder (for making 1X wash buffer), EIA plate reader

Patented technique uses a truly non-invasive method of detecting H. pylori antigens in human stool samples

  • CLIA Complexity: Non-Waived
  • CPT Code: 87338
  • Sensitivity: 96.1%
  • Specificity: 95.7%
  • Assay Format: ELISA
  • Break-away microtiter strips
  • Color-coded, ready-to-use reagents
  • No calculations required
  • Room temperature incubation
  • Qualitative test results in less than 90 minutes
  • For in vitro diagnostic use

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Clia Complexity Non-waived
Detectable Analytes H. pylori Antigen
Format Microwell
Sample Type Stool
Type Premier™ Platinum HpSA™ Plus
Includes Antibody coated microwells, positive control, negative control, sample diluent, 20X Wash Buffer, Enzyme conjugate, substrate, stop solution, transfer pipets, microwell strip holder, microwell strip sealer and wooden stick applicator, break-away microtiter strips
Quantity 96 Tests
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.