Learn More
MilliporeSigma™ Supelco™ Merlin MicroSeal System (fits Varian™) For CP-1177 injector
Catalog No. 111007799 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With:
Varian CP-1177 Injector
Catalog No. For Use With
11-100-7799 Varian CP-1177 Injector
1 options
Catalog No. 11-100-7799 Supplier MilliporeSigma™ Supelco™ Supplier No. 22609U
The Merlin Microseal System is a septum-less system that provides very long life.
The Merlin Microseal System is a septum-less system that provides very long life. Because there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner. The septum contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal. The septum can be used only with a syringe that has a 23 gauge 'blunt tipped' needle, or with an SPME fiber assembly with a 23 gauge needle. The thicker shaft is required to make the necessary contact with the septum seals.
TRUSTED_SUSTAINABILITY
Specifications
Includes 1 nut, 1 inlet adapter, 1 o-ring, and 1 General Purpose (3-100 psi) septum
For Use With Varian CP-1177 Injector
Varian is a registered trademark of Varian Medical Systems, Inc.