Learn More
MilliporeSigma™ Supelco™ Merlin MicroSeal™ System Replacement Septum 1 SPME septum
Catalog No. 111009767 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With:
SPME System
Catalog No. For Use With
11-100-9767 SPME System
1 options
Catalog No. 11-100-9767 Supplier MilliporeSigma™ Supelco™ Supplier No. 24818U
Add to Cart
Add to Cart
The Merlin MicroSeal™ System is a septum-less system that provides very long life.
The Merlin MicroSeal™ System is a septum-less system that provides very long life. Because there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner. The septum contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal. The septum can be used only with a syringe that has a 23 gauge 'blunt tipped' needle, or with an SPME fiber assembly with a 23 gauge needle. The thicker shaft is required to make the necessary contact with the septum seals.
TRUSTED_SUSTAINABILITY
Specifications
For Use With SPME System
MicroSeal is a trademark of Merlin Instrument Co.