Learn More
MilliporeSigma™ Supelco™ Merlin MicroSeal™ System Replacement Septum
Catalog No. 111009978 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Pressure:
1 to 45 psi
3 to 100 psi
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pressure
11-100-9978 1 to 45 psi
11-100-9762 3 to 100 psi
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9978 Supplier MilliporeSigma™ Supelco™ Supplier No. 22583
Add to Cart
Add to Cart
The Merlin MicroSeal™ System is a septum-less system that provides very long life.
The Merlin MicroSeal™ System is a septum-less system that provides very long life. Because there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner. The septum contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal. The septum can be used only with a syringe that has a 23 gauge 'blunt tipped' needle, or with an SPME fiber assembly with a 23 gauge needle. The thicker shaft is required to make the necessary contact with the septum seals.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chromatography Applications
Description 1 Low Pressure Septum
Pressure 1 to 45 psi
MicroSeal is a trademark of Merlin Instrument Co.