Learn More
Merlin Microseal™ System, Sold by MilliporeSigma™ Supelco™
Catalog No. 111008782 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
11-100-8782 1 Ea.
1 options
Catalog No. 11-100-8782 Supplier MilliporeSigma Supelco Supplier No. 22581U
Includes 1 Nut and 2 Low Pressure (1-45 psi) Septa, Fits Agilent GCs.
  • A septum-less system that provides very long life
  • Since there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner
  • Contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal
  • Can be used only with a syringe that has a 23 gauge blunt tipped needle, or with an SPME Fiber Assembly with a 23 gauge needle
  • A thicker shaft is required to make the necessary contact with the septum seals
TRUSTED_SUSTAINABILITY
Specifications
Description Includes 1 Nut and 2 Low Pressure (1-45 psi) Septa
For Use With (Equipment) Agilent GCs
For Use With (Application) Replacing Traditional Septa to Avoid Leaking and Bleeding
Quantity 1 Ea.
MicroSeal is a trademark of Merlin Instrument Co.