Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Meropenem Trihydrate, Beta-lactam Antibiotic, Solid, 10 mg

Catalog No. 502707074
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7074 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7074 Supplier Abcam Supplier No. AB12097510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Broad-spectrum beta-lactam antibiotic that is blood-brain barrier permeable.

  • Molecular weight: 437.51 g/mol
  • Purity: >98%
  • Description: Broad-spectrum β-lactam antibiotic
  • Form: Solid
  • Properties: Blood-brain barrier permeable
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Meropenem
Substrate Properties Chemical Substrate
Buffer 100% Meropenem trihydrate
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Formulation 100% Meropenem trihydrate
Quantity 10 mg
For Use With (Application) Antibiotic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.