Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Meropenem Trihydrate, Beta-Lactam Antibiotic, Solid, 50 mg

Catalog No. 502707073
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7073 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7073 Supplier Abcam Supplier No. AB12097550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Broad-spectrum beta-lactam antibiotic that is blood-brain barrier permeable.

  • Molecular weight: 437.51 g/mol
  • Purity: >98%
  • Description: Broad-spectrum β-lactam antibiotic
  • Characteristics: Blood-brain barrier permeable
  • Form: Solid
  • Composition: 100% Meropenem trihydrate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Meropenem trihydrate
Target Molecule Meropenem Trihydrate
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Meropenem trihydrate
Quantity 50 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.