Learn More
Abcam Mertansine (DM1 Compound), microtubule-targeted cytoto x ic agent, 50MG
Supplier:  Abcam AB14609650MG
Encompass_Preferred
MW 738.3 Da. Potent microtubule-targeted cytotoxic agent. Induces mitotic arrest. Inhibits microtubule polymerization. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3242