Learn More
Abcam Mertansine, Microtubule-targeted Cytotoxic Agent, Solid, 25 mg
Description
- Molecular Weight: 738.3 Da
- Form: Solid
- Mechanism of Action: Potent microtubule-targeted cytotoxic agent; induces mitotic arrest and inhibits microtubule polymerization
- Biological Activity: Shows antitumor effects in vivo
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Microtubules |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% N2’-deacetyl-N2’-(3-mercapto-1-oxopropyl) maytansine |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 25 mg |
| For Use With (Application) | Cancer research, antitumor studies, microtubule research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.