Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES, >98%, MP Biomedicals™

Catalog No. ICN19530983 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19530983 250 g
ICN19530910 10 g
ICN19530925 25 g
ICN19530980 100 g
ICN19530991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19530983 Supplier MP Biomedicals, Inc Supplier No. 0219530983
Add to Cart
Add to Cart
Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym 2-[N-Morpholino]ethanesulfonic acid
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range >98%
Formula Weight 213.2
Chemical Name or Material MES
EINECSNumber 249-259-4
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.