Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MES, Monohydrate, OmniPur™, Calbiochem™,

Catalog No. 61201KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-201-KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-201-KG Supplier MilliporeSigma™ Supplier No. 61201KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular biology grade buffer suitable for laboratory and manufacturing applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym mes monohydrate, 2-morpholinoethanesulfonic acid hydrate, mes hydrate, mes buffer hydrate, 4-morpholineethanesulfonic acid monohydrate, 4-morpholineethanesulfonic acid, monohydrate, 2-4-morpholinyl ethanesulfonic acid hydrate, mes hydrate buffer, 4-morpholineethanesulfonicacid, hydrate 1:1, mesmonohydrate
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Color White
Quantity 1 kg
Solubility Information Water: Soluble
Formula Weight 213.25
Percent Purity ≥99.0%
Grade Molecular biology grade
Physical Form Solid
Chemical Name or Material MES, Monohydrate
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.