Learn More
Cellular Engineering Technologies Mesenchymal Stem Cell Expansion Media (450mL)
Supplier:  Cellular Engineering Technologies MR1016
Encompass_Preferred
Media is specifically designed for high-density growth of mesenchymal stem cells and undifferentiated somatic stem cells. This media promotes robust growth and will maintain stem cells in an undifferentiated state. Media is shipped with gel packs.
Catalog No. NC2753090