Learn More
STEMCELL Technologies MesenCult™-hPL Medium Kit, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 05439
Encompass
MesenCult™-hPL Medium has been optimized for the culture and expansion of human mesenchymal stromal cells (MSCs; also known as mesencyhymal stem cells) as well as for the detection and enumeration of colony-forming unit fibroblasts (CFU-F). MesenCult™-hPL 10X Supplement is an alternative growth supplement to fetal bovine serum (FBS) that contains purified human platelet lysate (hPL). Complete MesenCult™-hPL Medium (Basal Medium + 10X Supplement) does not require addition of growth factors, lipids, or attachment substrate. It is more defined and provides better performance than FBS-containing media, and is also a more affordable option than defined, serum-free media.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. 50-197-4347
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.