Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-Butane-1,2,3,4-tetracarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B068425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B068425G 25 g
B0684500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B068425G Supplier TCI America Supplier No. B068425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4534-68-3
Molecular Formula C8H10O8
Molecular Weight (g/mol) 234.16
MDL Number MFCD00070496
InChI Key GGAUUQHSCNMCAU-ZXZARUISSA-N
PubChem CID 6992626
IUPAC Name (2S,3R)-butane-1,2,3,4-tetracarboxylic acid
SMILES C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 234.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material meso-Butane-1,2,3,4-tetracarboxylic Acid
EINECSNumber (2)-0897&(2)-1485
RTECSNumber EK6100000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.