Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-Tetraphenylchlorin, TCI America™
SDP

Catalog No. T1358100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1358100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1358100MG Supplier TCI America Supplier No. T1358100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2669-65-0
Molecular Formula C44H32N4
MDL Number MFCD00213944

Specifications

Color Blue-Purple
Quantity 100 mg
Formula Weight 616.77
Physical Form Crystalline Powder
Chemical Name or Material meso-Tetraphenylchlorin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.