Learn More
Diagnostic Biosystems MESOTHELIN Mesothelin
Supplier:  Diagnostic Biosystems RMAB107
Encompass_Preferred
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, which is present on the surface of normalmesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignantmesotheliomas.
Catalog No. 50-238-5520
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.