Learn More
US Biomax Inc MESOTHELIOMA ARRAY40CS/80 CORE
SDP
Supplier:  US Biomax Inc MS801B
Encompass
Mesothelioma array with mesothelium tissue, including TNM and clinical stage, 40 cases/80 cores, replacing MS801a
Catalog No. NC2097519
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.