Learn More
Invitrogen™ MESP1 Polyclonal Antibody

Description
MESP1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | MESP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MESP1 |
| Gene Accession No. | P97309 |
| Gene Alias | bHLHc5; Class C basic helix-loop-helix protein 5; mesoderm posterior 1; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; mesoderm posterior bHLH transcription factor 1; mesoderm posterior protein 1; MESP1; MGC10676 |
| Gene Symbols | MESP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.