Learn More
AdipoGen Mesuximide
SDP
Supplier:  AdipoGen CDXM0153M050
Encompass_Preferred
Chemical. CAS 77-41-8. Formula C12H13NO2. MW 203.24. Synthetic A succinimide with anti-epileptic and anti-convulsant properties. The mechanisms of action are unknown and possibly multiple. Metabolically reduced to its pharmacologically active metabolite N-desmethylmethosuximide.
Catalog No. 50-256-0492
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.