Learn More
Enzo Life Sciences Met-AMC (fluorogenic substrate) (25mg). CAS: 94367-35-8
SDP
Supplier:  Enzo Life Sciences BMLP2360025
Encompass_Preferred
Fluorogenic substrate for methionine aminopeptidase 2 (MetAP2) (kcat/Km=1.1 x 10 M-1min-1; Km=310 µM) or aminopeptidase N (Km=377 µM). Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Purity: ≥98% (TLC). Long Term Storage: -20°C.
Catalog No. 50-201-1303
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.