Learn More
CPC Scientific H-Tyr-Gly-Gly-Phe-Met-OH 100MG
SDP
Supplier:  CPC Scientific ENPK006C
Encompass
Opioid agonist that inhibits vas deferens contractions mediated through delta receptors.SEQUENCE: H-Tyr-Gly-Gly-Phe-Met-OHONE-LETTER SEQUENCE: YGGFMMOLECULAR FORMULA: C27H35N5O7S1MOLECULAR WEIGHT: 573.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [58569-55-4]SYNONYMS: Endorphin (1-5), Opioid Growth FactorRESEARCH AREA: PainREFERENCES:J. Hughes et al., Nature, 258, 577 (1975)SKU(s): ENKP-006A, ENKP-006B, ENPK-006C
Catalog No. 50-210-9260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.