Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MET Mouse anti-Human, Clone: UMAB190, liquid, UltraMAB™
SDP

Catalog No. 501685290
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5290 Supplier OriGene Supplier No. UM800082
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MET is a tyrosine kinase receptor for the hepatocyte growth factor. It is linked to functions such as cell proliferation, scattering, morphogenesis, and survival. Ligand binding at the cell surface induces autophosphorylation of MET that provides docking sites for downstream signaling molecules. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.

Specifications

Antigen MET
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UMAB190
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MET
Gene Accession No. P08581
Gene Alias AUTS9, DFNB97, HGFR, RCCP2, c-Met
Gene Symbols MET
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 570-885 of human MET produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4233
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.