Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Metal 25-Person Kit

Catalog No. 19090666 Shop All Medique Products
Change view
Click to view available options
Dimensions (L x W x H):
9.5 x 6.75 x 2.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
19-090-666 9.5 x 6.75 x 2.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-090-666 Supplier Medique Supplier No. 818ANSI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides first aid for up to 25 people for several emergency situations.

  • Wall-mounted
  • Easy to open and used on the go

Specifications

Product Type 25-Person Kit
For Use With (Application) For Emergency Situations
Material Metal
Sufficient For For 25 People
Dimensions (L x W x H) 9.5 x 6.75 x 2.75 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.