Learn More
Description
- Small, metal beaker tongs
- Rubber coated tips for slip free grip
Specifications
Specifications
| For Use With (Application) | Grasping Beakers with 25 - 100 mL Capacities |
| Tip Style | Rubber Coated |
| Product Type | Beaker Tongs |
| Material | Metal |
| Length (Metric) | 18.41 cm |
| Length (English) | 7.25 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
