Learn More
Mitegen Metal Cryo Grid Box, Aluminum, Holds 4 Autogrid/TEM Grids, Numbered Wells, Pin-Type Lid, Easy Tweezer Access
SDP
Supplier:  Mitegen MCEMAGG1001
Encompass
This Metal Cryo Grid Box provides easy access grid storage in cryogenic conditions. Manufactured from durable, shatter-proof cryo-grade aluminum, it offers a long lifespan, ensuring enhanced protection for delicate samples. The box features numbered wells (1-4) specifically made to accommodate autogrid rings and standard TEM grids, allowing easy tweezer access for precise manipulation. Its innovative pin-type lid incorporates a unique step, effectively preventing pick-up tools from sliding down or getting caught, enhancing handling security. Compatible with autogrid rings and widely used loading systems, this grid box also supports clipped grids and actively reduces electrostatic charging. It can be thoroughly cleaned with solvents or autoclaved to prevent cross-contamination, ensuring reusability. Optional custom serialization is available for enhanced sample tracking. For optimal handling, specialized gripper tools (SKUs M-CEM-AGT1003 or M-CEM-AGT1002) are recommended.
Catalog No. NC1816920
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.