Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Metal Inertia Ball with 3 Fixed Eye Bolts
SCI_ED

Catalog No. S12937 Shop All Eisco Products
Change view
Click to view available options
Metal Inertia Ball with 3 Fixed Eye Bolts; Weight: 455 g (1 lb.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S12937 Metal Inertia Ball with 3 Fixed Eye Bolts; Weight: 455 g (1 lb.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12937 Supplier Eisco™ Supplier No. PH0311
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suspend from the ceiling or a stand for multiple experiments. The Metal Inertia Ball with 3 Fixed Eye Bolts is used to demonstrate Newton's First Law of motion or can also be used as a pendulum bob.

  • Great for numerous inertia labs
  • Painted black
  • Heavy-duty 1 lb. metal ball
  • Fitted with 3 eye bolts
  • Eye bolts have 3/8 in. inside diameter

Specifications

Product Type Inertia Demonstration
Description Fitted with 3 eye bolts (3/8 in. D)
Weight (English) 1 lb.
Weight (Metric) 455 g
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.