Learn More
Eisco™ Metal Jolly Bulb
SCI_ED
Catalog No. S24872 Shop All Eisco Products
Change view
Click to view available options
Palm sized
Catalog No.
S24872 Palm sized
1 options
Catalog No. S24872 Supplier Eisco™ Supplier No. PH0138D
Add to Cart
Add to Cart
For demonstration of behavior of ideal gas and change of pressure.
  • Hollow metallic ball
  • Palm-sized
  • Manometer connected to investigate change of pressure in closed volume of air when heated or cooled
Specifications
Product Type Jolly Bob
Sensor Types Manometer
For Use With (Application) Exploring the relationship between pressure and temperature for an ideal gas
Includes Hollow metal sphere directly attached to a pressure gauge
Length (English) 7.75 in.