Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Metal Lysing Tube, 2 mL, with 0.75 x 0.5 in. Grinding Cylinder

Catalog No. MP116992002 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 Pk.
3 Pk.
6 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116992002 2 Pk.
MP116992003 3 Pk.
MP116992006 6 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP116992002 Supplier MP Biomedicals™ Supplier No. 116992002
Add to Cart
Add to Cart

Stainless steel lysing matrix tube with the grinding cylinder is ideal for grinding, lysis, and homogenization of resistant samples.

This stainless steel lysing matrix tube with the grinding cylinder is ideal for grinding, lysis, and homogenization of resistant samples. Constructed from 308 SS, these tubes and grinding cylinders are tough enough to stand up to the most demanding mechanical punishment that can cause traditional thermoplastic tubes to crack. The tubes are machined from premium grade billet and deliver superior strength over less expensive production methods such as deep-drawn aluminum tubes. An oblique angle conical bottom provides a better impact surface than the rounded bottoms of deep-drawn tubes. The stainless steel threaded cap provides a leak-proof closure without the energy-robbing alternatives like plastic flange screw caps or rubber stoppers. A Teflon O-ring prevents leakage, and can be cleaned with detergent and/or autoclaving, or replaced entirely between samples. Machined knurls on the cap provide a firm grip to easy opening and closing.

Sample types include:

  • Tough seeds such as dried corn, soybeans, wheat, tomato, and Chile
  • Wood, bark, roots, animal claws and hooves
  • Bone, teeth, hair, fingernails, and non-biological substrates
  • Tough tissues, bone, cartilage, skin
  • Non-biological, rocks and minerals, plastics and composites, printed circuit boards, wood, and building materials
Application Notes
  • Dry grinding of very tough or hard samples in which generated heat can damage plastic tubes.
  • Cryogenic dry grinding where severe cold temperatures (dry ice or liquid nitrogen) can damage plastic tubes.
  • Milling or grinding non-biological samples where plastic contamination is of concern.
  • Sample processing with solvents or chemicals that are incompatible with plastics.
TRUSTED_SUSTAINABILITY

Specifications

Bead Type Stainless Steel
Sample Type Seeds, Wood, Bark, Roots, Bone, Teeth, Tissues, Cartilage, Rocks, Minerals, Plastics, Composites
Includes 0.75 x 0.5 in. grinding cylinder
Product Type Metal Lysing Tube
Quantity 2 Pk.
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.