Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Metal Packed GC Column (fits Agilent, Configuration A), Phase None, Matrix 60/80 Carboxen-1000 Support

Catalog No. 111016207 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
4.6 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-101-6207 4.6 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6207 Supplier MilliporeSigma™ Supelco™ Supplier No. 12392U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An economical metal column should be used for less demanding applications where the inertness of glass is not required.

This column fits Agilent 5880, 5890, 6890, 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gas Chromatography (GC)
Length (Metric) 4.6 m
Material Stainless Steel Column
Packing Material Matrix 60/80 Carboxen-1000
Diameter (Metric) Inner 2.1 mm
Length (English) 15 ft.
Diameter (English) Outer 1/8 in.
Product Type Packed Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.