Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Metalguard™ 1880 Heavy Weight Gloves
GSA_VA

Catalog No. 17200382 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-382 Large
17-200-379 X-Small
17-200-380 Small
17-200-381 Medium
17-200-383 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-200-382 Supplier Wells Lamont™ Supplier No. 1880L
Add to Cart
Add to Cart
This item is not returnable. View return policy

High level cut-resistant Metalguard™ 1880 Heavy Weight Gloves feature ANSI Level A6 protection for use in metal handling and other applications.

  • Reinforced thumb crotch for longer life
  • Continuous knit for close secure fit
  • Ambidextrous
  • Prewashed and preshrunk
  • ANSI Level A6 cut resistance
  • Made in the USA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance ASTM F2992-15, ASTM D3884-09
Construction Seamless Knit
Coating Material Uncoated
Color Yellow, Green
Cut Resistance Rating ANSI A6
Hand Ambidextrous
Material Aramid Fiber, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Steel processing, metal handling, stamping, fabricating and framing
Glove Cuff Color White
Model 1880L
Series Metalguard™ 1880
Yarn Gauge 7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.