Learn More
Metals in Impinger Solution, Proficiency Testing Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Quantity | 250 mL |
| Concentration or Composition (by Analyte or Components) | Antimony, Sb 0.15-25 μg/mL; Arsenic, As 0.15-25 μg/mL; Barium, Ba 0.15-25 μg/mL; Beryllium, Be 0.15-25 μg/mL; Cadmium, Cd 0.15-25 μg/mL; Chromium, Cr(III) 0.15-25 μg/mL; Cobalt, Co 0.15-25 μg/mL; Copper, Cu 0.15-25 μg/mL; Lead, Pb 0.15-25 μg/mL; Manganese, Mn 0.15-25 μg/mL; Nickel, Ni 0.15-25 μg/mL; Selenium, Se 0.15-25 μg/mL; Silver, Ag 0.15-25 μg/mL; Thallium, Tl 0.15-25 μg/mL; Zinc, Zn 0.15-25 μg/mL |