Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Metamorphic Rock Set
SDPSCI_ED

Catalog No. S89550 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89550 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89550 Supplier United Scientific™ Supplier No. UNMETRC01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Learn about the transformation of existing rock types due to extreme heat and pressure with the United Scientific™ Metamorphic Rock Set

Learn about the transformation of existing rock types due to extreme heat and pressure with the United Scientific Metamorphic Rock Set. This process is called metamorphism, which means change in form.

  • The set includes anthracite, biotite schist, gneiss, marble black, marble green, marble pink, marble white, mica schist, phyllite, quartzite gray, quartzite green, quartzite white, serpentine, slate, tiger eye
  • Average sizes are 2 x 3 x 1.5 cm

Specifications

Grade 5 to 12
Format Kit
Product Type Rock Set
Includes 15 Specimens
Languages English
Subject Geology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.