Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 717200
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-720-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-720-0 Supplier Invitrogen™ Supplier No. 717200
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

METAP2 Polyclonal Antibody for Western Blot, ELISA

The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintenance of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METAP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene METAP2
Gene Accession No. O08663, P50579, Q3ZC89
Gene Alias 4930584B20Rik; A930035J23Rik; AI047573; AL024412; Amp2; AU014659; eIF-2-associated p67; eIF-2-associated p67 homolog; initiation factor 2 associated 67 kDa protein; initiation factor 2-associated 67 kDa glycoprotein; initiation factor 2-associated protein (p67); MAP 2; MAP2; metAP 2; Metap2; Methionine aminopeptidase 2; methionyl aminopeptidase 2; Mnpep; p67; p67eIF2; Peptidase M; peptidase M 2; similar to methionyl aminopeptidase 2; testicular tissue protein Li 17
Gene Symbols METAP2
Host Species Rabbit
Immunogen Fusion protein between the catalytic domain of human Methionine Aminopeptidase-2 (MetAP-2) and GST.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10988, 404150, 56307
Target Species Human, Mouse, Bovine
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.