Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METAP2 Recombinant Rabbit Monoclonal Antibody (JG39-79)
GREENER_CHOICE

Catalog No. PIMA534804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534804 Supplier Invitrogen™ Supplier No. MA534804
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Daudi, K562, mouse thymus tissue lysate, mouse kidney tissue, LOVO, PC-3M, SH-SY-5Y, rat seminal vesicle tissue, human tonsil tissue, human prostate cancer tissue, human kidney tissue, mouse testis tissue.

The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintenance of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METAP2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JG39-79
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene METAP2
Gene Accession No. P50579
Gene Alias 4930584B20Rik; A930035J23Rik; AI047573; AL024412; Amp2; AU014659; eIF-2-associated p67; eIF-2-associated p67 homolog; initiation factor 2 associated 67 kDa protein; initiation factor 2-associated 67 kDa glycoprotein; initiation factor 2-associated protein (p67); MAP 2; MAP2; metAP 2; Metap2; Methionine aminopeptidase 2; methionyl aminopeptidase 2; Mnpep; p67; p67eIF2; Peptidase M; peptidase M 2; similar to methionyl aminopeptidase 2; testicular tissue protein Li 17
Gene Symbols METAP2
Host Species Rabbit
Immunogen Recombinant protein within Human METAP2 aa 340-478.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10988
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.