Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Metapneumovirus Nucleoprotein Recombinant Rabbit Monoclonal Antibody (HL2921)
GREENER_CHOICE

Catalog No. MA556749
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA556749 Supplier Invitrogen™ Supplier No. MA556749
  • $522.00
    $391.50 / Each of 1

    Save $130.50

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Metapneumovirus Nucleoprotein Recombinant Monoclonal Antibody for Western Blot, ICC/IF, LF

The Metapneumovirus Nucleoprotein is a key structural protein of the human metapneumovirus (HMPV), a virus belonging to the Paramyxoviridae family that causes respiratory tract infections, particularly in young children, the elderly, and immunocompromised individuals. The nucleoprotein (N protein) encapsidates the viral RNA genome, forming a helical ribonucleoprotein complex that is essential for the replication and transcription of the viral RNA. This protein not only protects the viral RNA from degradation but also plays a crucial role in the assembly and release of new viral particles from infected cells. The N protein is highly conserved and immunogenic, making it a significant target for diagnostic assays and vaccine development. Understanding the structure and function of the metapneumovirus nucleoprotein is critical for developing effective antiviral therapies and for advancing our knowledge of HMPV pathogenesis and immune response mechanisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Metapneumovirus Nucleoprotein
Applications Lateral Flow, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2921
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias h h h-MPV; h-MPV; MPV
Host Species Rabbit
Immunogen Full length Human metapneumovirus Nucleoprotein. (strain CAN97-83).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.