Learn More
Impacto™ Metatarsal Protector
Impacto™ Metguard Metatarsal Protector protects the metatarsal area of the foot and fits all safety shoes and boots with optional attachment straps.
Supplier:  Impacto™ METGUARD
Catalog No. 19-400-037
Only null left
Add to Cart
Description
Description
Impacto™ Metguard Metatarsal Protector
  • Made of lightweight polycarbonate material
  • Secured with existing laces or optional straps
  • Non-conductive for heat and electricity
  • Used with existing footwear
  • Suitable for use in workplaces and industries including mining, construction, manufacturing, transport, storage, forestry, retail trade
  • Meets minimum impact resistance 75 lbs. as specified in ASTM F2413/F2412 (ANSI Z41)
  • Specifications
    Specifications
    Black
    Universal
    Unisex
    Metatarsal protector
    Polycarbonate
    Plain Toe
    Yes
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Promotions
    Promotions
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.