Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metaxalone 98.0+%, TCI America™
SDP

Catalog No. M25781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25781G 1 g
M25785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25781G Supplier TCI America Supplier No. M25781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1665-48-1
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.256
MDL Number MFCD00867700
InChI Key IMWZZHHPURKASS-UHFFFAOYSA-N
Synonym 5-(3,5-Dimethylphenoxymethyl)-2-oxazolidinone, 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone
PubChem CID 15459
ChEBI CHEBI:6797
IUPAC Name 5-[(3,5-dimethylphenoxy)methyl]-1,3-oxazolidin-2-one
SMILES CC1=CC(=CC(=C1)OCC2CNC(=O)O2)C

Specifications

Melting Point 124°C
Color White
Boiling Point 225°C
Quantity 1 g
Formula Weight 221.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Metaxalone
RTECSNumber RQ3850000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.