Learn More
Medchemexpress LLC HY-B0627 10mg Medchemexpress, Metformin CAS:657-24-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB062710MG
Encompass_Preferred
Medchemexpress, HY-B0627 10mg Metformin CAS:657-24-9 Metformin (1,1-Dimethylbiguanide) inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin can cross the blood-brain barrier and triggers autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1107
Due to product restrictions, please to purchase or view availability for this product.