Learn More
Selleck Chemical LLC Metformin
Supplier: Selleck Chemical LLC S595850MG
Metformin (1 1-Dimethylbiguanide) a widely used drug for treatment of type 2 diabetes activates AMP-activated protein kinase (AMPK) in hepatocytes Metformin promotes mitophagy in mononuclear cells Metformin induces apoptosis of lung cancer cells through activating JNK/p38 MAPK pathway and GADD153
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.