Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacholine chloride, 100.4%, MP Biomedicals™

Catalog No. ICN19023125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19023125 25 g
ICN19023105 5 g
ICN19023110 10 g
ICN19023180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19023125 Supplier MP Biomedicals, Inc Supplier No. 0219023125
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62-51-1
Molecular Formula C8H18ClNO2
Molecular Weight (g/mol) 195.687
MDL Number MFCD00011817
InChI Key JHPHVAVFUYTVCL-UHFFFAOYSA-M
Synonym methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride
PubChem CID 6114
ChEBI CHEBI:50142
IUPAC Name 2-acetyloxypropyl(trimethyl)azanium;chloride
SMILES CC(C[N+](C)(C)C)OC(=O)C.[Cl-]

Specifications

Color White
Assay Percent Range ≥98%
Quantity 25 g
Formula Weight 195.7
Physical Form Powder
Chemical Name or Material Methacholine chloride
Recommended Storage Store at 0°C.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.