Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Methacryloxypropyltrimethoxysilane, 99.04%, MP Biomedicals™

Catalog No. ICN15537805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15537805 5 mL
ICN15537825 25 mL
ICN15537850 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15537805 Supplier MP Biomedicals, Inc Supplier No. 0215537805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2530-85-0
Molecular Formula C10H20O5Si
Molecular Weight (g/mol) 248.35
MDL Number MFCD00008593
InChI Key XDLMVUHYZWKMMD-UHFFFAOYSA-N
Synonym 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
PubChem CID 17318
IUPAC Name 3-trimethoxysilylpropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC[Si](OC)(OC)OC

Specifications

Color Colorless
Density 1.04g/mL
Quantity 5 mL
Formula Weight 248.35
Percent Purity 0.9904
Physical Form Liquid
Chemical Name or Material γ-Methacryloxypropyl-trimethoxysilane, Adhesive
Recommended Storage Store at 4°C under nitrogen.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.