CAS RN 2530-85-0
3-(Trimethoxysilyl)propyl methacrylate, 98%
CAS: 2530-85-0 Molecular Formula: C10H20O5Si Molecular Weight (g/mol): 248.35 MDL Number: MFCD00008593 InChI Key: XDLMVUHYZWKMMD-UHFFFAOYSA-N Synonym: 3-trimethoxysilyl propyl methacrylate,3-methacryloxypropyltrimethoxysilane,methacryloxypropyltrimethoxysilane,dynasylan memo,silicone a-174,union carbide a-174,mops-m,silane a-174,nuca 174,2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester PubChem CID: 17318 IUPAC Name: 3-trimethoxysilylpropyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCCC[Si](OC)(OC)OC
γ-Methacryloxypropyltrimethoxysilane, 99.04%, MP Biomedicals™
CAS: 2530-85-0 Molecular Formula: C10H20O5Si Molecular Weight (g/mol): 248.35 MDL Number: MFCD00008593 InChI Key: XDLMVUHYZWKMMD-UHFFFAOYSA-N Synonym: 3-trimethoxysilyl propyl methacrylate,3-methacryloxypropyltrimethoxysilane,methacryloxypropyltrimethoxysilane,dynasylan memo,silicone a-174,union carbide a-174,mops-m,silane a-174,nuca 174,2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester PubChem CID: 17318 IUPAC Name: 3-trimethoxysilylpropyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCCC[Si](OC)(OC)OC
3-(Trimethoxysilyl)propyl Methacrylate (stabilized with BHT) 98.0+%, TCI America™
CAS: 2530-85-0 Molecular Formula: C10H20O5Si Molecular Weight (g/mol): 248.35 MDL Number: MFCD00008593 InChI Key: XDLMVUHYZWKMMD-UHFFFAOYSA-N Synonym: 3-trimethoxysilyl propyl methacrylate,3-methacryloxypropyltrimethoxysilane,methacryloxypropyltrimethoxysilane,dynasylan memo,silicone a-174,union carbide a-174,mops-m,silane a-174,nuca 174,2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester PubChem CID: 17318 IUPAC Name: 3-trimethoxysilylpropyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCCC[Si](OC)(OC)OC
Cytiva PlusOne™ Reagents
High quality electrophoresis reagents. Cytiva Life Sciences™ PlusOne Reagents are a low toxicity, environmentally safe solvent for high quality electrophoresis.
3-(Methacryloyloxy)propyltrimethoxysilane, 97%
CAS: 2530-85-0 Molecular Formula: C10H20O5Si Molecular Weight (g/mol): 248.35 MDL Number: MFCD00008593 InChI Key: XDLMVUHYZWKMMD-UHFFFAOYSA-N Synonym: 3-trimethoxysilyl propyl methacrylate,3-methacryloxypropyltrimethoxysilane,methacryloxypropyltrimethoxysilane,dynasylan memo,silicone a-174,union carbide a-174,mops-m,silane a-174,nuca 174,2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester PubChem CID: 17318 IUPAC Name: 3-trimethoxysilylpropyl 2-methylprop-2-enoate
Sigma Aldrich 3-(Trimethoxysilyl)propyl methacrylate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.