Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methallyltrimethylsilane 95.0+%, TCI America™
SDP

Catalog No. M301025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M301025ML 25 mL
M30105ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M301025ML Supplier TCI America Supplier No. M301025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18292-38-1
Molecular Formula C7H16Si
Molecular Weight (g/mol) 128.29
MDL Number MFCD00054404
InChI Key OWVJMAKUWHECNI-UHFFFAOYSA-N
Synonym Trimethyl(2-methylallyl)silane
PubChem CID 642554
IUPAC Name trimethyl(2-methylprop-2-enyl)silane
SMILES CC(=C)C[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 mL
UN Number 1993
Formula Weight 128.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methallyltrimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.