Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanesulfonic acid methyl ester, 99.4%, For GC analysis, MP Biomedicals

Catalog No. ICN15538425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15538425 25 g
ICN15538405 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15538425 Supplier MP Biomedicals, Inc Supplier No. 0215538425

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66-27-3
Molecular Formula C2H6O3S
Molecular Weight (g/mol) 110.127
InChI Key MBABOKRGFJTBAE-UHFFFAOYSA-N
Synonym methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate
PubChem CID 4156
ChEBI CHEBI:25255
IUPAC Name methyl methanesulfonate
SMILES COS(=O)(=O)C

Specifications

Color Colorless
Density 1.3g/mL
Quantity 25 g
Formula Weight 110.13
Percent Purity 0.994
Physical Form Liquid
Chemical Name or Material Methanesulfonic acid methyl ester, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.