Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol with 0.1% Acetic Acid, LabReady™ Solvent Blend, Solstice

Catalog No. 6000050 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-000-50 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-000-50 Supplier Solstice Supplier No. 4644
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-56-1 , 64-19-7
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point 68°C
CAS Max % 99.9000%, 0.1000%
UN Number UN1230
Density 0.79 g/cm3
Grade Solvent Blend
Packaging Glass Bottle
Quantity 4 x 4 L
Flash Point 20°C
Chemical Name or Material Methanol with 0.1% Acetic Acid, LabReady™ Solvent Blend
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.